Learn More
Essendant Stripping Floor Pads, 20"" Diameter, Black, 5/Carton
Catalog No. 502003461
Encompass_Preferred
For long-lasting stripping of finish, dirt and buildup. Use with rotary machines (Speed of 175-600 rpms). Color-coded for easy job identification.
Catalog No. 50-200-3461 Supplier Essendant Supplier No. BWK4020BLA
For long-lasting stripping of finish, dirt and buildup. Use with rotary machines (Speed of 175-600 rpms). Color-coded for easy job identification.