Learn More
Uline Tabletop Impulse Sealer with Cutter 20 Inch
Catalog No. NC0776134
Encompass
20 inch manual heat impulse bag sealer with built-in guillotine cutter for sealing poly bags and plastic sheeting up to 20 inches wide, adjustable temperature control for varying bag thicknesses, and Teflon coated sealing bars to prevent bag material sticking.
Catalog No. NC0776134 Supplier Uline Supplier No. H1252
20 inch manual heat impulse bag sealer with built-in guillotine cutter for sealing poly bags and plastic sheeting up to 20 inches wide, adjustable temperature control for varying bag thicknesses, and Teflon coated sealing bars to prevent bag material sticking.