Learn More
Alkali Scientific 2000mL (2L) Erlenmeyer Flask with Vented Cap and Flat Bottom, PC, Sterile, 4/cs
SDP
Catalog No. 502336004
Encompass_Preferred
Our Erlenmeyer Flasks are made from PETG with an HDPE cap and are manufactured in air filtered clean rooms. These Erlenmeyer Flasks are ideal for shaker culture applications. Erlenmeyer Flasks are sterilized by E-Beam and are non-pyrogenic, DNase and RNase free. They are non-toxic and environmentally friendly. Vent caps have 0.22 μm hydrophobic filters for gas exchange without contamination.
Catalog No. 50-233-6004 Supplier Alkali Scientific Supplier No. EFN607
Our Erlenmeyer Flasks are made from PETG with an HDPE cap and are manufactured in air filtered clean rooms. These Erlenmeyer Flasks are ideal for shaker culture applications. Erlenmeyer Flasks are sterilized by E-Beam and are non-pyrogenic, DNase and RNase free. They are non-toxic and environmentally friendly. Vent caps have 0.22 μm hydrophobic filters for gas exchange without contamination.