Learn More
Ace Glass, Inc. Mantle, fits 22 liter spherical flask, 350mm ID x 173mm depth, 115v 50/60 Hz, (2)770w circuits, CSA rated
SDP
Catalog No. 50977329
Encompass
20L aluminum housed, table-model mantle for Schott spherical flask. 350mm I.D. x 173mm height. Maximum internal operating temperature is 450 Deg. centigrade. (2)770w-115v, cULus rated. Power cord included.
Catalog No. 50-977-329 Supplier Ace Glass, Inc. Supplier No. 1205330
20L aluminum housed, table-model mantle for Schott spherical flask. 350mm I.D. x 173mm height. Maximum internal operating temperature is 450 Deg. centigrade. (2)770w-115v, cULus rated. Power cord included.