Learn More
Avantor Fluid Handling 20L Bag, Multi-layer EVA/LLDPE Barrier Film, 3-Ports, ADCF, 10/pk
Catalog No. NC0708731
Encompass
This item has a minimum order quantity of 20. BioProcess Bags from RIM Bio are optimized for specific container applications. RIM's bags provide maximum flex-cracking resistance, strength and durability and are well suited for cell culture applications. Each container product is constructed using NxFlex F1000C1 film, a medical grade multi-layer film specifically designed for high performance bioprocess applications. RIM utilizes both RF and Heat Impulse Sealing depending on the size and type of bag. The bags typically have Ports on one end and Handles on the opposing end. RIM offers two and three Port designs.
Catalog No. NC0708731 Supplier Avantor Fluid Handling Supplier No. RCBB2001
This item has a minimum order quantity of 20. BioProcess Bags from RIM Bio are optimized for specific container applications. RIM's bags provide maximum flex-cracking resistance, strength and durability and are well suited for cell culture applications. Each container product is constructed using NxFlex F1000C1 film, a medical grade multi-layer film specifically designed for high performance bioprocess applications. RIM utilizes both RF and Heat Impulse Sealing depending on the size and type of bag. The bags typically have Ports on one end and Handles on the opposing end. RIM offers two and three Port designs.