Learn More
Alkali Scientific 20L Carboy with Handles, Polypropylene (PP), TPE Gasket, Autoclavable, With Tap, 4/Case
SDP
Catalog No. 5000662128
Encompass_Preferred
This 20L Carboy with Handles is engineered for dependable liquid storage and dispensing in laboratory settings. Made from high quality, autoclavable materials, it ensures both durability and sterility, making it ideal for handling culture media, sterile water, and other lab solutions. Capacity: 20 liters, with molded volume markings for easy monitoring Material: High quality polypropylene (PP) for excellent chemical resistance Seal: Leak proof thermoplastic elastomer (TPE) gasket ensures a secure closure Sterilization: Fully autoclavable, including the barrel, cap, and tap Handles: Integrated side handles for safe and easy transport Design: Threaded cap and tap for controlled dispensing and tight sealing Applications: Suitable for storing and dispensing sterile water, buffer solutions, and culture media in lab, research, and pharmaceutical environments
Catalog No. 50-006-62128 Supplier Alkali Scientific Supplier No. CA20TCS
This 20L Carboy with Handles is engineered for dependable liquid storage and dispensing in laboratory settings. Made from high quality, autoclavable materials, it ensures both durability and sterility, making it ideal for handling culture media, sterile water, and other lab solutions. Capacity: 20 liters, with molded volume markings for easy monitoring Material: High quality polypropylene (PP) for excellent chemical resistance Seal: Leak proof thermoplastic elastomer (TPE) gasket ensures a secure closure Sterilization: Fully autoclavable, including the barrel, cap, and tap Handles: Integrated side handles for safe and easy transport Design: Threaded cap and tap for controlled dispensing and tight sealing Applications: Suitable for storing and dispensing sterile water, buffer solutions, and culture media in lab, research, and pharmaceutical environments