Learn More
Aqua Solutions Conductivity Standard (0.01 Molar)1,413 umhos @25°C (NIST) | 7447-40-7 | MFCD00011360 | 20L
SDP
Catalog No. NC1567432
Encompass_Preferred
Conductivity Standard (0.01 Molar)1,413 umhos @25°C (NIST) | 7447-40-7 | MFCD00011360 | 20L
Catalog No. NC1567432 Supplier Aqua Solutions Supplier No. 692020L
Conductivity Standard (0.01 Molar)1,413 umhos @25°C (NIST) | 7447-40-7 | MFCD00011360 | 20L