Learn More
Alkali Scientific 20L Digital Water Bath, bath water heater
SDP
Catalog No. 5000663039
Encompass_Preferred
This equipment is designed to handle a broad spectrum of laboratory procedures, including incubation, inactivation, and agglutination, as well as various pharmaceutical, serological, biomedical, and industrial processes. To ensure optimal temperature uniformity, the Water Bath is equipped with an energy efficient, large area heater and thermostatic control. Its PID microprocessor control system (Proportional Integral Derivative) offers precise heat control by anticipating the approach to your set point temperature and preventing overshoot. A redundant safety thermostat is standard on all models. Display: 3.25" LCD Working Temperature Range: Ambient +5 °C to 100 °C Temperature Stability: ±0.1 °C Temperature Calibration Capability: 1 point Working Access (L x W x D): 24.1 x 43.2 x 15.2 cm Flammability Class (DIN 12876 1): I (NFL)
Catalog No. 50-006-63039 Supplier Alkali Scientific Supplier No. LS20A11
This equipment is designed to handle a broad spectrum of laboratory procedures, including incubation, inactivation, and agglutination, as well as various pharmaceutical, serological, biomedical, and industrial processes. To ensure optimal temperature uniformity, the Water Bath is equipped with an energy efficient, large area heater and thermostatic control. Its PID microprocessor control system (Proportional Integral Derivative) offers precise heat control by anticipating the approach to your set point temperature and preventing overshoot. A redundant safety thermostat is standard on all models. Display: 3.25" LCD Working Temperature Range: Ambient +5 °C to 100 °C Temperature Stability: ±0.1 °C Temperature Calibration Capability: 1 point Working Access (L x W x D): 24.1 x 43.2 x 15.2 cm Flammability Class (DIN 12876 1): I (NFL)