Learn More
Chemglass Life Sciences 20L (Short) Insulating Jacket
SDP
Catalog No. CG10011120
Encompass_Preferred
20L (Short) Insulating Jacket. Similar to CG-10009 but without the heating element. For use as a heat or cold shield on Chemglass jacketed reactors. Insulating jacket is made from a heavy fiberglass fabric on the inside with a silicone-coated fiberglass fabric on the outside. Mantle is slit lengthwise to a hole in the bottom center with lacing closure to accommodate a bottom drain valve.
Catalog No. CG10011120 Supplier Chemglass Life Sciences Supplier No. CG10011120
20L (Short) Insulating Jacket. Similar to CG-10009 but without the heating element. For use as a heat or cold shield on Chemglass jacketed reactors. Insulating jacket is made from a heavy fiberglass fabric on the inside with a silicone-coated fiberglass fabric on the outside. Mantle is slit lengthwise to a hole in the bottom center with lacing closure to accommodate a bottom drain valve.