Learn More
Foxx Life Sciences EZBio Single Use Carboy (Bottle) Assembly, 20L (5GAL) Gamma Sterilized Polycarbonate (PC) Carboy for Aseptic Transfer and Sterile Storage with VersaCap 83B (83mm), Vent and Dip Tube, 1/EA
GREENER_CHOICE
Catalog No. NC1796841
Encompass_Preferred
EZBio Gamma Sterilized Carboys are with distinctive rectangular shape saves valuable bench space while the molded grips provide a secure hold to facilitate in lifting, handling, and pouring. With easy to read metric graduation marks, certified to 5% accuracy, these carboys include pre-mounted, high tensile strength, platinum-cured silicone tubing. This assembly provides a flexible two-port filling and venting system, allowing for extractable free, aseptic gas and liquid transfer as required for pharmaceutical, biotechnology, and laboratory fluid transfer applications. These high-quality carboy assemblies are readily able to accept additional fluid transfer components such as aseptic connectors, filters, and manifold integration. The optional wide neck allows users to reach inside the carboy for easier cleaning. Gamma Irradiated in the United States.AutoclavableUSP Class VI, FDA Grade materialsClear for high visibility of product
Catalog No. NC1796841 Supplier Foxx Life Sciences Supplier No. 558HE01FLS
EZBio Gamma Sterilized Carboys are with distinctive rectangular shape saves valuable bench space while the molded grips provide a secure hold to facilitate in lifting, handling, and pouring. With easy to read metric graduation marks, certified to 5% accuracy, these carboys include pre-mounted, high tensile strength, platinum-cured silicone tubing. This assembly provides a flexible two-port filling and venting system, allowing for extractable free, aseptic gas and liquid transfer as required for pharmaceutical, biotechnology, and laboratory fluid transfer applications. These high-quality carboy assemblies are readily able to accept additional fluid transfer components such as aseptic connectors, filters, and manifold integration. The optional wide neck allows users to reach inside the carboy for easier cleaning. Gamma Irradiated in the United States.AutoclavableUSP Class VI, FDA Grade materialsClear for high visibility of product