Learn More
Perkin Elmer US LLC 20µL Stainless Steel Sample Loop for Model 7725 RheodyneInjectors
Catalog No. 502104547
Encompass_Preferred
The size designations of stainless steel loops are nominal. Theactual volumes can differ from the theoretical designations becauseof the 0.025 mm (0.001) tolerance of the metal tubingbore. Accuracy of large metal loops (1.0 mm, 0.040 bore) is about5%, intermediate loops (0.5 mm, 0.020 bore) 10%,and small loops (0.2 mm, 0.007 bore) 30%. Bore: 0.30mm.Model 7725 Injector loops are not interchangeable with loops forthe Model 7125. The port angle for the 7725 is 30° whereas theport angle for the 7125 is 20°, requiring the loops to have adifferent shape.
Catalog No. 50-210-4547 Supplier Perkin Elmer US LLC Supplier No. N9306025
The size designations of stainless steel loops are nominal. Theactual volumes can differ from the theoretical designations becauseof the 0.025 mm (0.001) tolerance of the metal tubingbore. Accuracy of large metal loops (1.0 mm, 0.040 bore) is about5%, intermediate loops (0.5 mm, 0.020 bore) 10%,and small loops (0.2 mm, 0.007 bore) 30%. Bore: 0.30mm.Model 7725 Injector loops are not interchangeable with loops forthe Model 7125. The port angle for the 7725 is 30° whereas theport angle for the 7125 is 20°, requiring the loops to have adifferent shape.