Learn More
S2 Media 20mL 6x Lauryl Tryptose Broth (LTB-6 or 6xLSB) with Bromocresol Purple in 125mL Media Bottle, 1/each
Catalog No. 502995923
Encompass_Preferred
S2Media™ 6x Lauryl Tryptose Broth (LTB-6 or 6xLSB) with Bromocresol Purple is a selective liquid medium ued for the detection of coliforms in materials of sanitary importance. The fermentation of lactose with gas formation, typically collected in an inverted Durham tube, is a presumptive test for coliforms. Bromocresol purple is a pH indicator, causing the solution to shift from purple to yellow when lactose fermentation occurs. The fermentation of lactose with gas formation, typically collected in an inverted Durham tube, is a presumptive test for coliforms. Sodium lauryl sulfate inhibits non-coliform organisms. • Product meets AOAC, APHA, BAM, EPA, and SMWW reference methods and performance criteria.• 6xLSB should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known was Lauryl Sulfate Tryptose (LST) Broth or Lauryl Sulfate Broth (LSB).
Catalog No. 50-299-5923 Supplier S2 Media Supplier No. 5352
S2Media™ 6x Lauryl Tryptose Broth (LTB-6 or 6xLSB) with Bromocresol Purple is a selective liquid medium ued for the detection of coliforms in materials of sanitary importance. The fermentation of lactose with gas formation, typically collected in an inverted Durham tube, is a presumptive test for coliforms. Bromocresol purple is a pH indicator, causing the solution to shift from purple to yellow when lactose fermentation occurs. The fermentation of lactose with gas formation, typically collected in an inverted Durham tube, is a presumptive test for coliforms. Sodium lauryl sulfate inhibits non-coliform organisms. • Product meets AOAC, APHA, BAM, EPA, and SMWW reference methods and performance criteria.• 6xLSB should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known was Lauryl Sulfate Tryptose (LST) Broth or Lauryl Sulfate Broth (LSB).