Learn More
Research Products International Corp 20 ml Aluminum Weighing Pans, 100 per Package
SDP
Catalog No. 50998221
Encompass
Disposable round aluminum dishes can be used as an evaporating dish, weighing dish, parts container, dust cover and more. They are oil residue free without a vinyl coating. Crimped sides provide rigidity and a sturdy gauge for heavy duty use. Their large tabs provide space for marking with sample numbers. The flat bottom improves sample handling and the sides are angled for easy separation.
Catalog No. 50-998-221 Supplier Research Products International Corp Supplier No. 144300
Disposable round aluminum dishes can be used as an evaporating dish, weighing dish, parts container, dust cover and more. They are oil residue free without a vinyl coating. Crimped sides provide rigidity and a sturdy gauge for heavy duty use. Their large tabs provide space for marking with sample numbers. The flat bottom improves sample handling and the sides are angled for easy separation.