Learn More
Alkali Scientific ASI™ 20mL Borosilicate Glass Scintillation Vials (27.5x57.5mm) – 0.67 oz., with White Polyethylene Caps (22-400 Thread), PE Liner Septa, 500/Case
SDP
Catalog No. 503028651
Encompass_Preferred
ASI™ 20mL Borosilicate Glass Scintillation Vials are designed for high-performance liquid scintillation counting, sample storage, and general laboratory applications. Made from premium borosilicate glass, these vials offer excellent chemical resistance, minimal background count, and superior clarity. Each vial features a 22-400 threaded finish and comes with a secure, leak-resistant white polyethylene cap for optimal sample protection. Capacity: 20mL (0.67 oz.) Dimensions: 27.5mm diameter x 57.5mm height Material: Borosilicate glass for superior chemical and thermal resistance Cap: White polyethylene screw cap with 22-400 thread for a tight seal Applications: Ideal for liquid scintillation counting, sample containment, and laboratory storage Packaging: 100 vials per pack Ensure sample integrity and reliable performance with ASI™ 20mL Scintillation Vials, trusted by laboratories for high-precision applications.
Catalog No. 50-302-8651 Supplier Alkali Scientific Supplier No. SCNT020PECS
ASI™ 20mL Borosilicate Glass Scintillation Vials are designed for high-performance liquid scintillation counting, sample storage, and general laboratory applications. Made from premium borosilicate glass, these vials offer excellent chemical resistance, minimal background count, and superior clarity. Each vial features a 22-400 threaded finish and comes with a secure, leak-resistant white polyethylene cap for optimal sample protection. Capacity: 20mL (0.67 oz.) Dimensions: 27.5mm diameter x 57.5mm height Material: Borosilicate glass for superior chemical and thermal resistance Cap: White polyethylene screw cap with 22-400 thread for a tight seal Applications: Ideal for liquid scintillation counting, sample containment, and laboratory storage Packaging: 100 vials per pack Ensure sample integrity and reliable performance with ASI™ 20mL Scintillation Vials, trusted by laboratories for high-precision applications.