Learn More
WELCH MATERIALS INC DoprahTM 20mL 27.5x57mm Clear Glass EPA/TOC Vial 24-400 White Open Top PP Screw Cap with 22mm Natural PTFE/White Silicone 3.0mm thick Septa (EPA Quality) and Dust Cover. 100pcs/pk.
Catalog No. NC2078204
Encompass_Preferred
20mL 27.5x57mm Clear Glass EPA/TOC Vial 24-400 White Open Top PP Screw Cap with 22mm Natural PTFE/White Silicone 3.0mm thick Septa (EPA Quality) and Dust Cover. 100pcs/pk.
Catalog No. NC2078204 Supplier WELCH MATERIALS INC Supplier No. DK1338
20mL 27.5x57mm Clear Glass EPA/TOC Vial 24-400 White Open Top PP Screw Cap with 22mm Natural PTFE/White Silicone 3.0mm thick Septa (EPA Quality) and Dust Cover. 100pcs/pk.