Learn More
Agilent Technologies 20ML CLEAR SCR TOP VIAL 100/PK
Catalog No. NC2227626
Encompass
Vial, screw top, headspace, clear, 20 mL, 23 x 75 mm, 100/pk. Vial size: 22.75 x 75 mm (18 mm cap), 100 per pack
This item is not returnable. View return policy
Catalog No. NC2227626 Supplier Agilent Technologies Supplier No. 51882753
Vial, screw top, headspace, clear, 20 mL, 23 x 75 mm, 100/pk. Vial size: 22.75 x 75 mm (18 mm cap), 100 per pack