Learn More
Chemglass Life Sciences Weighing Dish, Plastic, Square, Antistatic, 7mL, 41 x 41 x 8mm,Polystyrene
SDP
Catalog No. 501215162
Encompass_Preferred
The polystyrene weighing dishes will resist diluted acids, aqueous solutions, alcohols and bases. They are ideal for weighing, dispensing or storing both liquid and powdered samples. They afford accurate pour-outs with minimal sample loss and can also be used as quick freeze trays for sample material. Dishes will withstand temperatures up to 80°C.
Catalog No. 50-121-5162 Supplier Chemglass Life Sciences Supplier No. CLS1815001
The polystyrene weighing dishes will resist diluted acids, aqueous solutions, alcohols and bases. They are ideal for weighing, dispensing or storing both liquid and powdered samples. They afford accurate pour-outs with minimal sample loss and can also be used as quick freeze trays for sample material. Dishes will withstand temperatures up to 80°C.