Learn More
Ace Glass, Inc. Sample flask, 20ml, 19/22 top and bottom, 7/15 siphon tube
SDP
Catalog No. 50978543
Encompass_Preferred
20mL capacity neutral oil determination chromatography reservoir has 19/22 standard taper joints at the top and bottom, and a 7/15 standard taper joint on the inner tube. Meets A.O.C.S. CA9F-57 specifications.
Catalog No. 50-978-543 Supplier Ace Glass, Inc. Supplier No. 586908
20mL capacity neutral oil determination chromatography reservoir has 19/22 standard taper joints at the top and bottom, and a 7/15 standard taper joint on the inner tube. Meets A.O.C.S. CA9F-57 specifications.