Learn More
Alkali Scientific 20mL Glass Crimp Top Headspace
SDP
Catalog No. 503027443
Encompass_Preferred
Designed for everyday chromatography analysis ASI20 mL Glass Crimp Top Headspace Vials offer an affordable high-quality solution compatible with all headspace GC instruments Manufactured from clear Type I borosilicate glass these vials provide exceptional thermal and chemical resistance making them ideal for volatile gas analysis Key Features Superior Glass Quality Made from 1st hydrolytic class borosilicate glass (Type I) for enhanced chemical resistance and durability Beveled Finish Reduces the sealing surface between the crimp seal and the glass for a more secure and consistent closure Standard 20 mm Crimp Top Ensures compatibility with a wide range of crimp seals and autosamplers Optimal Size for Headspace Analysis 22 5 75 mm dimensions allow precise sample containment Thicker Glass Walls Designed to withstand higher temperatures and internal pressures ensuring reliable performance in headspace GC applications Specification Certified Dimensionally verified and inspec
Catalog No. 50-302-7443 Supplier Alkali Scientific Supplier No. GC020RB
Designed for everyday chromatography analysis ASI20 mL Glass Crimp Top Headspace Vials offer an affordable high-quality solution compatible with all headspace GC instruments Manufactured from clear Type I borosilicate glass these vials provide exceptional thermal and chemical resistance making them ideal for volatile gas analysis Key Features Superior Glass Quality Made from 1st hydrolytic class borosilicate glass (Type I) for enhanced chemical resistance and durability Beveled Finish Reduces the sealing surface between the crimp seal and the glass for a more secure and consistent closure Standard 20 mm Crimp Top Ensures compatibility with a wide range of crimp seals and autosamplers Optimal Size for Headspace Analysis 22 5 75 mm dimensions allow precise sample containment Thicker Glass Walls Designed to withstand higher temperatures and internal pressures ensuring reliable performance in headspace GC applications Specification Certified Dimensionally verified and inspec