Learn More
Chemglass Life Sciences 20ml Glass Scintillation Vials W/ 22-400 Black Phenolic Cone Lined Caps, 500/Case
SDP
Catalog No. 5000980665
Encompass_Preferred
Scintillation vials are made with low potassium borosilicate, Type 1 glass and will fit all liquid scintillation equipment.  Vials are packed into shrink wrapped compartmentalized trays of 100 pieces.
Catalog No. 50-009-80665 Supplier Chemglass Life Sciences Supplier No. CG494041
Scintillation vials are made with low potassium borosilicate, Type 1 glass and will fit all liquid scintillation equipment.  Vials are packed into shrink wrapped compartmentalized trays of 100 pieces.