Learn More
Chemglass Life Sciences 20ml Glass Scintillation Vials Without Caps, 22-400 Finish, 100/tray
SDP
Catalog No. 5000980696
Encompass_Preferred
Scintillation vials are made with low potassium borosilicate, Type 1 glass and will fit all liquid scintillation equipment. Vials are packed into shrink wrapped compartmentalized trays of 100 pieces.
Catalog No. 50-009-80696 Supplier Chemglass Life Sciences Supplier No. CG494010
Scintillation vials are made with low potassium borosilicate, Type 1 glass and will fit all liquid scintillation equipment. Vials are packed into shrink wrapped compartmentalized trays of 100 pieces.