Learn More
Chemglass Life Sciences 20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, Green Open Top Cap, TFE Septa, Clear\ PETG Tray
SDP
Catalog No. 5000451037
Encompass_Preferred
20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, Green Open Top Cap, TFE Septa, Clear\ PETG Tray. Graduated vials with marking spot, 1mL subdivisions from 5mL to 15mL on 20mL vials and 5mL to 30mL on 40mL vials. Packaged in a clear PETG tray complete with a slide or lift off dust cover. Vials are separated by a SBS partition to eliminate glass to glass contact. Caps are supplied compete in the package, not attached. Entire package is recyclable.Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware.Polypropylene caps have a septa bonded to the cap, using the Chemglass SURE-LINK™ process. This inert process bonds the .125” PTFE Faced Silicone septa directly to the cap without adhesives,
Catalog No. 50-004-51037 Supplier Chemglass Life Sciences Supplier No. CG4904G01
20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, Green Open Top Cap, TFE Septa, Clear\ PETG Tray. Graduated vials with marking spot, 1mL subdivisions from 5mL to 15mL on 20mL vials and 5mL to 30mL on 40mL vials. Packaged in a clear PETG tray complete with a slide or lift off dust cover. Vials are separated by a SBS partition to eliminate glass to glass contact. Caps are supplied compete in the package, not attached. Entire package is recyclable.Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware.Polypropylene caps have a septa bonded to the cap, using the Chemglass SURE-LINK™ process. This inert process bonds the .125” PTFE Faced Silicone septa directly to the cap without adhesives,