Learn More
Chemglass Life Sciences 20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, Green Open Top Cap, TFE Septa, SBS Coated White Tray
SDP
Catalog No. 5000451049
Encompass_Preferred
20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, Green Open Top Cap, TFE Septa, SBS Coated White Tray. Graduated vials with marking spot, 1mL subdivisions from 5mL to 15mL on 20mL vials and 5mL to 30mL on 40mL vials. Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identifi cation. Caps are supplied installed on the vials.
Catalog No. 50-004-51049 Supplier Chemglass Life Sciences Supplier No. CG4908G01
20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, Green Open Top Cap, TFE Septa, SBS Coated White Tray. Graduated vials with marking spot, 1mL subdivisions from 5mL to 15mL on 20mL vials and 5mL to 30mL on 40mL vials. Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware. For storage applications, these vials are packaged in a SBS coated white tray complete with SBS partition. Boxes are marked with alpha-numeric coordinates for fast and easy vial identifi cation. Caps are supplied installed on the vials.