Learn More
Chemglass Life Sciences 20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, Vial Only, LAB-PAC
SDP
Catalog No. 5000451043
Encompass_Preferred
20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, Vial Only, LAB-PAC. Graduated vials with marking spot, 1mL subdivisions from 5mL to 15mL on 20mL vials and 5mL to 30mL on 40mL vials packaged in convenient LAB-PACS. Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware. Vials are conveniently packed in shrink-wrapped packs of 100. Caps sold separately.
Catalog No. 50-004-51043 Supplier Chemglass Life Sciences Supplier No. CG4902G06
20mL Graduated Vials with Marking Spot, 28mm OD x 57mm Height, 24-400 Thread Size, Vial Only, LAB-PAC. Graduated vials with marking spot, 1mL subdivisions from 5mL to 15mL on 20mL vials and 5mL to 30mL on 40mL vials packaged in convenient LAB-PACS. Vials are made from 33 expansion borosilicate glass which conforms to ASTM E-438 Type 1-Class A, USP Type 1 and ISO 3585. The high resistance to thermal shock along with its maximum chemical resistance makes this the glass of choice for vials along with most all laboratory glassware. Vials are conveniently packed in shrink-wrapped packs of 100. Caps sold separately.