Learn More
SCP Science Fusion Crucible, Cylind., 20ml HP (10), 20 ml high purity graphite fusion cricibles with only 2 ppm of total metallic impurities | 10/pk
Catalog No. NC9496438
Encompass
Fusion Crucible, Cylind., 20ml HP (10), 20 ml high purity graphite fusion cricibles with only 2 ppm of total metallic impurities | 10/pk
Catalog No. NC9496438 Supplier SCP Science Supplier No. 040060031
Fusion Crucible, Cylind., 20ml HP (10), 20 ml high purity graphite fusion cricibles with only 2 ppm of total metallic impurities | 10/pk