Learn More
Alkali Scientific 20mL HDPE Scintillation Vials, Liquid Sample Scintillation Counting Vials, 22 400 White Screw Cap, Silver Aluminum Foil/Polyethylene Liner, 100/Pk
SDP
Catalog No. 5000662941
Encompass_Preferred
HDPE scintillation vials are designed to provide low background counts and excellent light transmission, making them ideal for assays that use reagents emitting low energy radiation. Resistant to swelling caused by solvent degradation, these vials ensure reliable performance in demanding laboratory applications. Each vial has a 20 mL capacity with dimensions of 27.5 × 57.5 mm and is paired with polypropylene caps that fit GPI 22 400 thread finishes, which can also be marked for easy sample tracking. For added convenience, the vials and caps are packaged in cellular trays with alphanumeric coordinates that allow quick and efficient identification. Used with scintillation counters, these vials facilitate the detection of light produced when phosphors in the cocktail are excited by radiation, supporting accurate and efficient measurement in scintillation assays.
Catalog No. 50-006-62941 Supplier Alkali Scientific Supplier No. SCNT020FHDPE
HDPE scintillation vials are designed to provide low background counts and excellent light transmission, making them ideal for assays that use reagents emitting low energy radiation. Resistant to swelling caused by solvent degradation, these vials ensure reliable performance in demanding laboratory applications. Each vial has a 20 mL capacity with dimensions of 27.5 × 57.5 mm and is paired with polypropylene caps that fit GPI 22 400 thread finishes, which can also be marked for easy sample tracking. For added convenience, the vials and caps are packaged in cellular trays with alphanumeric coordinates that allow quick and efficient identification. Used with scintillation counters, these vials facilitate the detection of light produced when phosphors in the cocktail are excited by radiation, supporting accurate and efficient measurement in scintillation assays.