Learn More
Trajan Scientific Chromatography, Autosampler Vials Caps and Closures, Vial, Headspace, Crimp, 20mL, 23x75mm, Round bottom, Case-1000
Catalog No. NC1803879
Encompass_Preferred
Chromatography, Autosampler Vials Caps and Closures, Vial, Headspace, Crimp, 20mL, 23x75mm, Round bottom, Case-1000
Catalog No. NC1803879 Supplier Trajan Scientific Supplier No. LAP.2009087310
Chromatography, Autosampler Vials Caps and Closures, Vial, Headspace, Crimp, 20mL, 23x75mm, Round bottom, Case-1000