Learn More
MOLTOX Molecular Toxicology Inc MUTAZYME 10%, Phenobarbital/5, 6-Benzoflavone induced S9 Mix, Lyophilized, Reconstitute to 20 mL, Store in Freezer
SDP
Catalog No. NC1323360
Encompass
Aroclor-1254 or Phenobarbital/5, 6-Benzoflavone induced Sprague Dawley male rat liver S9 lyophilized with NADPH-regenerating system cofactors and phosphate buffer. To use, simply reconstitute with the labeled volume of ice-cold, sterile, purified water. Available as 5%, 10% and 30% S9 Mix. Freeze dried using a proprietary process that confers exceptional stability. Especially useful where an ultra-low freezer is not available and where dry ice shipments are difficult or prohibitively expensive. Ready for use after reconstitution with the labeled volume of cold, sterile, purified water. MOLTOX S9 preparations and cofactor reagents for use in metabolic activation studies are manufactured to rigorous standards of quality and performance and are used by leading industrial, government and academic laboratories worldwide. *Non Returnable*
Catalog No. NC1323360 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 11404L
Aroclor-1254 or Phenobarbital/5, 6-Benzoflavone induced Sprague Dawley male rat liver S9 lyophilized with NADPH-regenerating system cofactors and phosphate buffer. To use, simply reconstitute with the labeled volume of ice-cold, sterile, purified water. Available as 5%, 10% and 30% S9 Mix. Freeze dried using a proprietary process that confers exceptional stability. Especially useful where an ultra-low freezer is not available and where dry ice shipments are difficult or prohibitively expensive. Ready for use after reconstitution with the labeled volume of cold, sterile, purified water. MOLTOX S9 preparations and cofactor reagents for use in metabolic activation studies are manufactured to rigorous standards of quality and performance and are used by leading industrial, government and academic laboratories worldwide. *Non Returnable*