Learn More
Chemglass Life Sciences Chemglass 20 mL Glass Scintillation Vial with 22-400 White Urea Cone-Lined Cap (CG-4940-30)
SDP
Catalog No. NC3352510
Encompass_Preferred
Chemglass 20 mL glass scintillation vials with a 22-400 screw finish and white urea cone-lined caps are designed for liquid scintillation counting and routine laboratory sample storage. The vials are made from low-potassium borosilicate (Type 1) glass for low background and chemical resistance. They are supplied in compartmentalized trays and shrink-wrapped for secure handling.
  • 20 mL capacity.
  • Low-potassium borosilicate (Type 1) glass.
  • 22-400 screw finish.
  • White urea cone-lined screw cap.
  • Packaged 100 per tray, shrink-wrapped compartmentalized trays.
  • Compatible with liquid scintillation equipment.
Catalog No. NC3352510 Supplier Chemglass Life Sciences Supplier No. CG494030
Chemglass 20 mL glass scintillation vials with a 22-400 screw finish and white urea cone-lined caps are designed for liquid scintillation counting and routine laboratory sample storage. The vials are made from low-potassium borosilicate (Type 1) glass for low background and chemical resistance. They are supplied in compartmentalized trays and shrink-wrapped for secure handling.
  • 20 mL capacity.
  • Low-potassium borosilicate (Type 1) glass.
  • 22-400 screw finish.
  • White urea cone-lined screw cap.
  • Packaged 100 per tray, shrink-wrapped compartmentalized trays.
  • Compatible with liquid scintillation equipment.