Learn More
Gerstel Screw Cap Vials, 20 mL, Clear, 100/Pack
SDP
Catalog No. NC9589986
Encompass
20 mL clear screw cap vials for MPS (rounded bottom for transport), Package of 100 Requires magnetic screw caps with septa 093640-092-00 (PolyMag) or 093640-040-00 (metal)
Catalog No. NC9589986 Supplier Gerstel Supplier No. 09364003600
20 mL clear screw cap vials for MPS (rounded bottom for transport), Package of 100 Requires magnetic screw caps with septa 093640-092-00 (PolyMag) or 093640-040-00 (metal)