Learn More
Chemglass Life Sciences Tube, Vacuum Hydrolysis, 18mL Capacity, 19mm Tube OD x 100mm Tube Length
SDP
Catalog No. CG402504
Encompass_Preferred
Tube, Vacuum Hydrolysis, 20mL Capacity, 19mm Tube OD x 100mm Tube Length. Tube is useful in protein hydrolysis, amino acid separations, freeze drying or any application under vacuum.
Catalog No. CG402504 Supplier Chemglass Life Sciences Supplier No. CG402504
Tube, Vacuum Hydrolysis, 20mL Capacity, 19mm Tube OD x 100mm Tube Length. Tube is useful in protein hydrolysis, amino acid separations, freeze drying or any application under vacuum.