Learn More
Analytical Sales & Services 20mm, 10mL, 23 x 46mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Flat Bottom
Catalog No. 5000806493
Encompass
Advantage™ 20mm, 10mL Volume, 23 x 46mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Flat Bottom
Catalog No. 50-008-06493 Supplier Analytical Sales & Services Supplier No. 621013
Advantage™ 20mm, 10mL Volume, 23 x 46mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Flat Bottom