Learn More
Analytical Sales & Services 20mm, 10mL, 23 x 46mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Round Bottom
Catalog No. 5000804384
Encompass
Advantage™ 20mm, 10mL Volume, 23 x 46mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Round Bottom - for PE Autsamplers
Catalog No. 50-008-04384 Supplier Analytical Sales & Services Supplier No. 672103
Advantage™ 20mm, 10mL Volume, 23 x 46mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Round Bottom - for PE Autsamplers