Learn More
Analytical Sales & Services 20mm, 20mL, 23 x 75mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Flat Bottom
Catalog No. 5000806555
Encompass
Advantage™ 20mm, 20mL Volume, 23 x 75mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Flat Bottom
Catalog No. 50-008-06555 Supplier Analytical Sales & Services Supplier No. 622013
Advantage™ 20mm, 20mL Volume, 23 x 75mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Flat Bottom