Learn More
Analytical Sales & Services 20mm, 6mL, 22 x 38mm Clear Glass Crimp Head Space Vial with Beveled Top and Beveled Bottom
Catalog No. 5000806310
Encompass
20mm, 6mL, 22 x 38mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Beveled Bottom
Catalog No. 50-008-06310 Supplier Analytical Sales & Services Supplier No. 622823
20mm, 6mL, 22 x 38mm Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Beveled Bottom