Learn More
Chemglass Life Sciences 20mm Aluminum Seals with PTFE/Silicone Septa
SDP
Catalog No. 50872811
Encompass_Preferred
20mm Aluminum Seals with PTFE/Silicone Septa. The 20mm Silver Aluminum Seals are supplied with 0.125" thick Blue PTFE/White Silicone Septa installed.
Catalog No. 50-872-811 Supplier Chemglass Life Sciences Supplier No. CG492010
20mm Aluminum Seals with PTFE/Silicone Septa. The 20mm Silver Aluminum Seals are supplied with 0.125" thick Blue PTFE/White Silicone Septa installed.