Learn More
Analytical Sales & Services 20mm Amber Glass Head Space Crimp Vial with Beveled Top and Flat Bottom
Catalog No. 5000804351
Encompass
Advantage™ 20mm Amber Glass Head Space Crimp Vial with Beveled Top and Flat Bottom\n\n• Amber Glass, Crimp Style\n• Inside Diameter of Opening: 20mm\n• Volume: 20mL\n• Outside Dimensions: 23mm x 75mm\n• Beveled Top, Flat Bottom\n• For PE Autsamplers\n\nQty/Pkg: 100
Catalog No. 50-008-04351 Supplier Analytical Sales & Services Supplier No. 672203
Advantage™ 20mm Amber Glass Head Space Crimp Vial with Beveled Top and Flat Bottom\n\n• Amber Glass, Crimp Style\n• Inside Diameter of Opening: 20mm\n• Volume: 20mL\n• Outside Dimensions: 23mm x 75mm\n• Beveled Top, Flat Bottom\n• For PE Autsamplers\n\nQty/Pkg: 100