Learn More
Analytical Sales & Services 20mm Blue Magnetic Aluminum/Tin Caps with Ultra Low Bleed Beige PTFE/White Silicone Liners
Catalog No. 5000806680
Encompass
Advantage™ 20mm Blue Magnetic Aluminum/Tin Caps with Ultra Low Bleed Beige PTFE/White Silicone Liners
Catalog No. 50-008-06680 Supplier Analytical Sales & Services Supplier No. 62800
Advantage™ 20mm Blue Magnetic Aluminum/Tin Caps with Ultra Low Bleed Beige PTFE/White Silicone Liners