Learn More
Chemglass Life Sciences 20mm Control Knob Only
SDP
Catalog No. 50873765
Encompass_Preferred
20mm Control Knob Only. Replacement 20mm Chem-Cap stem and control knob for Chemglass cylindrical and spherical vessels. PTFE stem is sold complete with Perfluoro or Viton o-rings installed under each Chem-Cap and one installed as the back up o-ring.
Catalog No. 50-873-765 Supplier Chemglass Life Sciences Supplier No. CG51131
20mm Control Knob Only. Replacement 20mm Chem-Cap stem and control knob for Chemglass cylindrical and spherical vessels. PTFE stem is sold complete with Perfluoro or Viton o-rings installed under each Chem-Cap and one installed as the back up o-ring.