Learn More
Perkin Elmer US LLC 20mm decapper jaw set for High Performance Crimper tool
Catalog No. 502111815
Encompass_Preferred
Decapper Jaw set for the High Performance Crimper. It fits standard 20 mm caps, is suitable for all steel and magnetic caps, and a wide range of seal thicknesses.
Catalog No. 50-211-1815 Supplier Perkin Elmer US LLC Supplier No. N9304512
Decapper Jaw set for the High Performance Crimper. It fits standard 20 mm caps, is suitable for all steel and magnetic caps, and a wide range of seal thicknesses.