Learn More
PATHOLOGY INNOVATIONS The Peters Precision Cryoembedding System 20x20 Deep Well Bar Set includes one 20x20mm Deep Well Bar, which has two 6mm deep wells and two 9mm deep wells, as well as 4 small chucks, two over chuck freezing blocks and 8 dispensing slides.
Catalog No. NC3370383
Encompass_Preferred
The Peters Precision Cryoembedding System is a combination of simple techniques and apparatus which allows the user to embed tissues for frozen section with a level of control and predictability that can surpass even paraffin embedding. The 20x20 Deep Well Bar Set includes one 20x20mm Deep Well Bar, which has two 6mm deep wells and two 9mm deep wells, as well as 4 small chucks, two over chuck freezing blocks and 8 dispensing slides. This system was designed by inventor, professor and author of frozen section literature Dr. Stephen Peters to give users more precision and control when doing a high volume of frozen section procedures. This set is made for outfitting labs that process a high volume of frozen sections with average sized specimen, and a great way to get started using the Peters Precision Cryoembedding System which has been adopted by thousands of hospitals globally. Learn how to use the system as well as advanced frozen section technique at pathologyinnovations.com.
Catalog No. NC3370383 Supplier PATHOLOGY INNOVATIONS Supplier No. 116
The Peters Precision Cryoembedding System is a combination of simple techniques and apparatus which allows the user to embed tissues for frozen section with a level of control and predictability that can surpass even paraffin embedding. The 20x20 Deep Well Bar Set includes one 20x20mm Deep Well Bar, which has two 6mm deep wells and two 9mm deep wells, as well as 4 small chucks, two over chuck freezing blocks and 8 dispensing slides. This system was designed by inventor, professor and author of frozen section literature Dr. Stephen Peters to give users more precision and control when doing a high volume of frozen section procedures. This set is made for outfitting labs that process a high volume of frozen sections with average sized specimen, and a great way to get started using the Peters Precision Cryoembedding System which has been adopted by thousands of hospitals globally. Learn how to use the system as well as advanced frozen section technique at pathologyinnovations.com.