Learn More
Chemglass Life Sciences 20mm Extended Tip Drain Valve, Complete
SDP
Catalog No. 501531786
Encompass_Preferred
20mm Extended Tip Drain Valve, Complete. Designed to be used as a drain for reaction kettles, flasks, etc. Valve is made with an extended tip which fills the seat area and permits total agitation of the contents of the vessel. Once opened, contents of the vessel will drain through the sidearm. Valve is constructed the same as the rest of Chemglass's Chem-Cap valve line, and has an integral PTFE tip o-ring. Supplied complete PTFE stem with Viton o-rings, control knob, and corresponding glass barrel having a single sidearm which can be further fabricated into the drain arm.
Catalog No. 50-153-1786 Supplier Chemglass Life Sciences Supplier No. CG51102
20mm Extended Tip Drain Valve, Complete. Designed to be used as a drain for reaction kettles, flasks, etc. Valve is made with an extended tip which fills the seat area and permits total agitation of the contents of the vessel. Once opened, contents of the vessel will drain through the sidearm. Valve is constructed the same as the rest of Chemglass's Chem-Cap valve line, and has an integral PTFE tip o-ring. Supplied complete PTFE stem with Viton o-rings, control knob, and corresponding glass barrel having a single sidearm which can be further fabricated into the drain arm.