Learn More
Azzota Corp 20mm Pathlength Standardfluorometer Cuvettes 7Ml - Pathlength 20mm - Inside Width 10mm Volume 7.0Ml Dimensions 45X12.5X22.5mm; Cover And Lid, Glass (340Nm-2500Nm)
Catalog No. 507532889
Encompass_Preferred
20mm Pathlength StandardFluorometer Cuvettes 7ml - Pathlength 20mm - Inside width 10mm Volume 7.0ml Dimensions 45x12.5x22.5mm; Cover and Lid, Glass (340nm-2500nm)
Catalog No. 50-753-2889 Supplier Azzota Corp Supplier No. G205
20mm Pathlength StandardFluorometer Cuvettes 7ml - Pathlength 20mm - Inside width 10mm Volume 7.0ml Dimensions 45x12.5x22.5mm; Cover and Lid, Glass (340nm-2500nm)