Learn More
Azzota Corp 20mm Pathlength Standard Cuvettes - 7Ml Pathlength - 20mm Inside Width - 10mm Volume - 7.0Ml Dimensions - 45X12.5X22.5mm Cover And Round Bottom Optical Glass (340Nm-2500Nm)
Catalog No. 507532883
Encompass_Preferred
20mm Pathlength Standard Cuvettes - 7ml Pathlength - 20mm Inside width - 10mm Volume - 7.0ml Dimensions - 45x12.5x22.5mm Cover and round bottom Optical Glass (340nm-2500nm)
Catalog No. 50-753-2883 Supplier Azzota Corp Supplier No. G105
20mm Pathlength Standard Cuvettes - 7ml Pathlength - 20mm Inside width - 10mm Volume - 7.0ml Dimensions - 45x12.5x22.5mm Cover and round bottom Optical Glass (340nm-2500nm)