Learn More
Ace Glass, Inc. 20MM GRADED SEALS 20MM GRADED SEALS
SDP
Supplier:  Ace Glass, Inc. 845532
Encompass
20mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-979-783