Learn More
Perkin Elmer US LLC 20 mm Hand Crimper
Catalog No. 509045917
Encompass_Preferred
For use on GC Headspace Vials or LC Large Volume Vials. Crimp capping is still a preferred method by some laboratories to ensure sample integrity, particularly those containing volatile compounds.
Catalog No. 50-904-5917 Supplier Perkin Elmer US LLC Supplier No. N9302785
For use on GC Headspace Vials or LC Large Volume Vials. Crimp capping is still a preferred method by some laboratories to ensure sample integrity, particularly those containing volatile compounds.