Learn More
West Pharmaceutical Services Inc 20mm NovaPure® Ready-to-Sterilize Bromobutyl 4023/50 Serum Stoppers, Qty. 3,000 stoppers/bag
Catalog No. NC3931016
Encompass
NovaPure® serum stoppers are made from 4023/50 a premium quality bromobutyl elastomer with West’s lowest particulate specification available. Laminated with FluroTec™ barrier film to help reduce particulate, limit drug interaction with the elastomer and reduce leachables. These components are provided washed (ready-to-sterilize) and 100% vision inspected. This stopper configuration can be used with European blowback, North American blowback, or straight wall (no blowback) vials. West item number: 70025047
Catalog No. NC3931016 Supplier West Pharmaceutical Services Inc Supplier No. 70025047
NovaPure® serum stoppers are made from 4023/50 a premium quality bromobutyl elastomer with West’s lowest particulate specification available. Laminated with FluroTec™ barrier film to help reduce particulate, limit drug interaction with the elastomer and reduce leachables. These components are provided washed (ready-to-sterilize) and 100% vision inspected. This stopper configuration can be used with European blowback, North American blowback, or straight wall (no blowback) vials. West item number: 70025047