Learn More
Azzota Corp 20mm Path Fluorometer 7.0ml Qz
Catalog No. 5000887034
Encompass_Preferred
45x22 5x12 5mm 20mm Pathlength Standard Fluorimeter Cuvettes 7 0ml Quartz 190nm-2500nm Four polished windows
Catalog No. 50-008-87034 Supplier Azzota Corp Supplier No. Q205
45x22 5x12 5mm 20mm Pathlength Standard Fluorimeter Cuvettes 7 0ml Quartz 190nm-2500nm Four polished windows