Learn More
Ace Glass, Inc. 20MM POR B TUBES 20MM POR B TUBES
SDP
Catalog No. 50974653
Encompass_Preferred
20mm filtering tube with porosity B (70-100 micron) fritted disc sealed on bottom. Overall length is 125mm.
Catalog No. 50-974-653 Supplier Ace Glass, Inc. Supplier No. 720914
20mm filtering tube with porosity B (70-100 micron) fritted disc sealed on bottom. Overall length is 125mm.