Learn More
Ace Glass, Inc. 20MM POR C TUBES 20MM POR C TUBES
SDP
Catalog No. 50974654
Encompass_Preferred
20mm filtering tube with porosity C (25-50 micron) fritted disc sealed on bottom. Overall length is 125mm.
Catalog No. 50-974-654 Supplier Ace Glass, Inc. Supplier No. 720916
20mm filtering tube with porosity C (25-50 micron) fritted disc sealed on bottom. Overall length is 125mm.